missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical

Catalog No. 18606231
Change view
Click to view available options
Quantity:
25 g
100 g
125 g
500 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606231 25 g Amber Glass Bottle
18606232 100 g Amber Glass Bottle
18606233 125 g Amber Glass Bottle
18606234 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606231 Supplier Spectrum Chemical Mfg Cor Supplier No. S108525GM

S1085, 7761-88-8, AgNO3

Spectrum™ Chemical Silver Nitrate, Crystal, Reagent, ACS is an inorganic compound with the formula AgNO3. Silver nitrate is an extremely versatile analytical laboratory reagent for both the qualitative and quantitative determination of halides, carbonates, hydroxides, sulfides and phosphates. Spectrum provides solid Reagent ACS grade silver nitrate for preparation of analytical laboratory reagent solutions. As an ACS grade Reagent, Spectrum Chemical manufactured Silver Nitrate is used as the quality reagent in standard testing to which other substances are graded. It has met the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle