missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate concentrate, For 1L standard solution, 0.1 M AgNO3 (0.1N), Solstice

Catalog No. 6001339
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001339 1 Pc.
60046732 6 x 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001339 Supplier Solstice Supplier No. 383101EA

for 1L standard solution, 0.1 M AgNO3 (0.1N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Quantity 1 Pc.
Linear Formula AgNO3
UN Number UN3264
Solubility Information Soluble in Water
IUPAC Name silver(1+) nitrate
Formula Weight 169.87g/mol
Grade Reagent
Odor Odorless
Packaging Ampoule
Appearance Colorless Liquid
Chemical Name or Material Silver nitrate concentrate
Show More Show Less
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338