Learn More
Silver Nitrate, Certified, 0.500N ±0.005N (0.5M), LabChem™
Supplier: LabChem LC227302
Specifications
| Silver Nitrate | |
| Colorless | |
| Liquid | |
| AgNO3 | |
| MFCD00003414 | |
| Soluble in water | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| CHEBI:32130 | |
| Certified | |
| Passes Test | |
| 1g/mL | |
| 1g/mL |
| 7761-88-8,7732-18-5 | |
| 91.51,8.49 | |
| 1 L | |
| AgNO3 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 24470 | |
| 169.87 | |
| 0.500N ±0.005N (0.5M) | |
| Amber Glass | |
| Nitrogen oxides; Thermal decomposition generates: Corrosive vapors |
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Do not breathe mist, spray, vapors.
Avoid release to the environment.
Wear eye protection, protective clothing, protective gloves, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature