missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, Certified, 0.0100N ±0.0002N (0.01M), LabChem™

Supplier:  LabChem LC225904

 View more versions of this product

Catalog No. LC225904

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Silver Nitrate
Colorless
Liquid
AgNO3
MFCD00003414
Soluble in water
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
Certified
Passes Test
1g/mL
1g/mL
7761-88-8,7732-18-5
99.83,0.17
4 L
AgNO3
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
169.87
0.0100N ±0.0002N (0.01M)
Amber Glass
Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions