missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, ACS Reagent, ≥99.0%, Honeywell Fluka™

Catalog No. 60045182
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Poly bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045182 25 g Glass Bottle
60045181 100 g Glass Bottle
6001318 500 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60045182 Supplier Honeywell Chemicals Supplier No. 20913925G
Only null left

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Melting Point 212°C (decomposition)(literature)
CAS Max % ≤100.0000%
Quantity 25 g
Linear Formula AgNO3
UN Number UN1493
IUPAC Name silver;nitrate
Formula Weight 169.87
Percent Purity >99%
Grade ACS Reagent
Packaging Glass Bottle
Density 4.350 g/cm3; Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver nitrate, ACS Reagent, ≥99.0%
Show More Show Less
Hazard Statement P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H290-H314-H410
ShelfLife 900 days from date of manufacture
DOTInformation Transport Hazard Class: 5.1; Packing Group:3: II; Proper Shipping Name: Silver nitrate
EINECSNumber 231-853-9
RTECSNumber VW4725000
Recommended Storage Room Temp