missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, ACS Reagent, ≥99.0%, Honeywell Fluka™

ACS Reagent,≥99.0%

Supplier:  Honeywell-Fluka 209139100G

 View more versions of this product

Catalog No. 60045181

Only null left
Explore available promotions

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Silver nitrate
ACS Reagent, ≥99.0%
≤100.0000%
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
>99%
Glass Bottle
212°C (decomposition)(literature)
7761-88-8
100 g
AgNO3
UN1493
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver;nitrate
24470
169.87
ACS Reagent
4.350 g/cm3; Vapor Density: 5.8 (vs air)

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
[Ag+].[O-][N+]([O-])=O
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P220-P260-P280-P305 + P351 + P338-P370 + P378

H272-H290-H314-H410

ShelfLife : 900 days from date of manufacture

DOTInformation : Transport Hazard Class: 5.1; Packing Group:3: II; Proper Shipping Name: Silver nitrate

EINECSNumber : 231-853-9

RTECSNumber : VW4725000

Recommended Storage : Room Temp

SDS
Documents

Documents

Promotions

Promotions