missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate ACS MP Biomedicals

Catalog No. ICN15257125
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15257125 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15257125 Supplier MP Biomedicals Inc Supplier No. 0215257125
Only null left

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver;nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Quantity 25 g
Formula Weight 169.87
Grade ACS
Density 4.35g/cm3
Chemical Name or Material Silver Nitrate