missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, 99+%, ACS reagent

Catalog No. AC419350500
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419350500 50 g Glass bottle
AC419351000 100 g Glass bottle
AC419352500 250 g Glass bottle
AC419355000 500 g Glass bottle
AC419350025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC419350500 Supplier Thermo Scientific Chemicals Supplier No. 419350500

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 444.0°C
Melting Point 212.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 50 g
Assay Percent Range 99+%
Linear Formula AgNO3
Fieser 01,1008; 02,366; 03,252; 04,429; 05,582; 07,321; 09,411; 10,350; 11,268; 12,257; 14,350; 15,39
Merck Index 15, 8657
Solubility Information Solubility in water: 219 g/100mL (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone and ether
Formula Weight 169.87
Percent Purity ≥99%
Grade ACS Reagent
Packaging Glass bottle
Decomposition Information >444°C
Density 4.35
Chemical Name or Material Silver nitrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep/Store away from clothing/ combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off im
EINECSNumber 231-853-9

RUO – Research Use Only