missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, 99.5%, for analysis

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

Supplier:  Thermo Scientific Chemicals 419361000

 View more versions of this product

Catalog No. AC419361000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications

Specifications

444.0°C
212.0°C
Green to Red
100 g
AgNO3
MFCD00003414
15, 8657
Solubility in water: 219 g/100mL (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone and ether
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
99.5%
Passes Test
>444°C
Silver nitrate
7761-88-8
Crystalline Powder and/or Crystals
99.5%
AgNO3
01,1008; 02,366; 03,252; 04,429; 05,582; 07,321; 09,411; 10,350; 11,268; 12,257; 14,350; 15,39
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
169.87
Analytical
Glass bottle
4.35

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 231-853-9

RTECSNumber : VW4725000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only