missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, (1mL = 10mg NaCl), Certified, 0.171N ±0.002N (0.171M), LabChem™

Supplier:  LabChem LC227205

 View more versions of this product

Catalog No. LC227205

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Silver Nitrate
7761-88-8,7732-18-5
97.1,2.9
20 L
AgNO3
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
169.87
0.171N ±0.002N (0.171M)
Cubitainer™
Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide
(1mL = 10mg NaCl)
Colorless
Liquid
AgNO3
MFCD00003414
Soluble in water
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
Certified
Passes Test
1g/mL
1g/mL

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation and serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions