missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, (1mL = 10mg NaCl), Certified, 0.171N ±0.002N (0.171M), LabChem™
Supplier: LabChem LC227204
Specifications
| Silver Nitrate, 0.171N (0.171M) | |
| 7761-88-8,7732-18-5 | |
| 97.1,2.9 | |
| 4 L | |
| AgNO3 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 24470 | |
| 169.87 | |
| 0.171N ± 0.002N | |
| Amber Glass | |
| Nitrogen oxides; fume; Carbon monoxide; Carbon dioxide |
| 1mL = 10mg NaCl | |
| Colorless | |
| Liquid | |
| AgNO3 | |
| MFCD00003414 | |
| Soluble in water | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| CHEBI:32130 | |
| Certified | |
| Passes Test | |
| 1g/mL | |
| 1g/mL |
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
Causes skin irritation and serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before
Warning
Recommended Storage : Room Temperature