missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, 1.0N Standardized Solution

Catalog No. p-4407816
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35597AP 500 mL
AA35597K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35597AP Supplier Thermo Scientific Chemicals Supplier No. 035597AP
Only null left

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

Silver nitrate, 1.0N Standardized Solution is used for volumetric analysis in laboratories. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
Physical Form Liquid
Quantity 500 mL
Solubility Information Miscible with water,alcohol and aqueous ammonia.
Formula Weight 169.87
Sensitivity Light sensitive
Appearance Colorless liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Chemical Name or Material Silver nitrate
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packing Group: II
EINECSNumber 231-853-9
RTECSNumber VW4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only