missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate, 0.200 N (N/5), Ricca Chemical
Supplier: Ricca Chemical Company 70055
| Quantity | 20 L |
|---|---|
| Packaging | Cubitainer |
Chemical Identifiers
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
Specifications
| Silver Nitrate | |
| 95.0, 3.22 | |
| AgNO3 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| CHEBI:32130 | |
| 20 L |