missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.200 N (N/5), Ricca Chemical

Supplier:  Ricca Chemical Company 70051

Quantity 4 L
Packaging Amber Glass Bottle

 Afficher plus de versions de ce produit

Numéro de catalogue. 70051

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

Spécifications

Identifiants chimiques

7761-88-8 , 7732-18-5
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O

Spécifications

Silver Nitrate
95.0, 3.22
AgNO3
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
4 L
7761-88-8 , 7732-18-5
99.0, 3.36
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
Amber Glass Bottle
0.200Normal
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions