missing translation for 'onlineSavingsMsg'
Learn More

Silver(II) Pyridine-2-carboxylate 97.0+%, TCI America™

Catalog No. S08151G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08151G 1 g
S08155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08151G Supplier TCI America Supplier No. S08151G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14783-00-7 | C12H8AgN2O4-2 | 352.074 g/mol

Chemical Identifiers

CAS 14783-00-7
Molecular Formula C12H8AgN2O4-2
Molecular Weight (g/mol) 352.074
InChI Key ARXVTYGOLNYKET-UHFFFAOYSA-L
Synonym Picolinic Acid Silver(II) Salt, Silver(II) Picolinate, Pyridine-2-carboxylic Acid Silver(II) Salt
PubChem CID 13898161
IUPAC Name pyridine-2-carboxylate;silver
SMILES C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].[Ag]

Specifications

Quantity 1 g
Formula Weight 352.07
Percent Purity ≥97.0% (T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Silver(II) Pyridine-2-carboxylate
TSCA No
Recommended Storage Refrigerator