accessibility menu, dialog, popup

UserName

NOONOOAg

CAS RN 14783-00-7

IUPAC Name: bis(pyridine-2-carboxylate) silver
Synonyms: dipicolinate silver
Molecular Weight (g/mol): 352.08
Molecular Formula: C12H8AgN2O4
InChi Key: ARXVTYGOLNYKET-UHFFFAOYSA-L
SMILES: [Ag].[O-]C(=O)C1=CC=CC=N1.[O-]C(=O)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results