missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) chromate, 96%

Catalog No. p-7059783
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4549506 5 g
AA4549522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4549506 Supplier Thermo Scientific Chemicals Supplier No. 04549506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7784-01-2 | Ag2CrO4 | 331.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7784-01-2
Molecular Formula Ag2CrO4
Molecular Weight (g/mol) 331.73
MDL Number MFCD00003402
InChI Key OJKANDGLELGDHV-UHFFFAOYSA-N
Synonym silver chromate, unii-zj5092lwu9, silver chromate vi, silberchromat, silver i chromate, 2ag.cro4, chromic acid h2cro4 , disilver 1+ salt, chromic acid h2cro4 , silver 1+ salt 1:2, chromic acid, disilver i salt, disilver dioxido dioxo chromium
PubChem CID 62666
IUPAC Name disilver;dioxido(dioxo)chromium
SMILES [O-][Cr](=O)(=O)[O-].[Ag+].[Ag+]

Specifications

Color Red to Brown
Physical Form Powder
Quantity 5 g
Linear Formula Ag2CrO4
UN Number UN1479
Merck Index 14,8510
Formula Weight 331.73
Percent Purity 99.9%
Odor Odorless
Sensitivity Light sensitive
Density 5.63 g/mL
Assay (metals basis)
Chemical Name or Material Silver (I) chromate
Show More Show Less
Hazard Category H272-H317-H350i
Hazard Statement GHS H Statement
H350-H272-H317
May cause cancer.
May intensify fire
oxidizer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P220-P221-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber 232-043-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only