accessibility menu, dialog, popup

UserName

AgAgOCrOOO

CAS RN 7784-01-2

IUPAC Name: disilver(1+) dioxochromiumbis(olate)
Synonyms: silver chromate
Molecular Weight (g/mol): 331.73
Molecular Formula: Ag2CrO4
InChi Key: OJKANDGLELGDHV-UHFFFAOYSA-N
SMILES: [Ag+].[Ag+].[O-][Cr]([O-])(=O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results