missing translation for 'onlineSavingsMsg'
Learn More

Silver Cyanide, Powder, Purified, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1060125GM

 View more versions of this product

Catalog No. 18613129


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

506-64-9
125 g
Purified
1
99%
Amber Glass Bottle

Chemical Identifiers

506-64-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.