accessibility menu, dialog, popup

UserName

AgCN

CAS RN 506-64-9

IUPAC Name: silver(1+) iminomethanide
Synonyms: silver cyanide
Molecular Weight (g/mol): 133.89
Molecular Formula: CAgN
InChi Key: LFAGQMCIGQNPJG-UHFFFAOYSA-N
SMILES: [Ag+].[C-]#N
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (4)
Physical Form 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Grade 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (3)
  • (2)
  • (1)
  • (4)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)