missing translation for 'onlineSavingsMsg'
Learn More

Silver Cyanide, Powder, Purified, 99%, Spectrum™ Chemical

Catalog No. 18613127
Change view
Click to view available options
Quantity:
25 g
100 g
125 g
500 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613127 25 g Amber Glass Bottle
18613128 100 g Amber Glass Bottle
18613129 125 g Amber Glass Bottle
18613130 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613127 Supplier Spectrum Chemical Mfg Cor Supplier No. S106025GM

Chemical Identifiers

CAS 506-64-9

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade Purified
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.