missing translation for 'onlineSavingsMsg'
Learn More

Silver cyanide, 99%

Catalog No. AA1142414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1142414 25 g
AA1142422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1142414 Supplier Thermo Scientific Chemicals Supplier No. 01142414

Silver plating | CAS: 506-64-9 | CAgN | 133.886 g/mol

Silver cyanide is used in silver-plating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 506-64-9
Molecular Formula CAgN
Molecular Weight (g/mol) 133.886
MDL Number MFCD00003409
InChI Key LFAGQMCIGQNPJG-UHFFFAOYSA-N
Synonym silver cyanide, silver cyanide ag cn, silver 1+ cyanide, kyanid stribrny czech, cyanure d'argent french, rcra waste number p104, silver i cyanide, rcra waste no. p104, kyanid stribrny, cyanure d'argent
PubChem CID 10475
IUPAC Name silver;cyanide
SMILES [C-]#N.[Ag+]

Specifications

Melting Point 320°C (decomposition)
Color White
Physical Form Powder
Quantity 25 g
Linear Formula AgCN
UN Number UN1684
Merck Index 14,8512
Solubility Information Insoluble in water,dilute acids,and alcohol; Soluble in alkali cyanides and boiling concentrated HNO3
Formula Weight 133.84
Percent Purity 99%
Odor Odorless
Refractive Index 1.685
Sensitivity Light sensitive
Density 3.95 g/mL
Chemical Name or Material Silver cyanide
Show More Show Less
Hazard Category H300+H310+H330
Hazard Statement GHS H Statement
H300-H310-H330-H318-H290-H315
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
May be corrosive to metals.
Causes skin irritation.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SILVER CYANIDE
EINECSNumber 208-048-6
RTECSNumber VW3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only