missing translation for 'onlineSavingsMsg'
Learn More

Silver bromide, Premion™, 99.998% (metals basis)

CAS: 7785-23-1 | AgBr | 187.772 g/mol

Supplier:  Thermo Scientific Chemicals 01211014

 View more versions of this product

Catalog No. AA1211014


Only null left
Explore available promotions

Description

Description

Silver bromide is used in photographic film. Used in production of radiograph. It also used in infrared applications as windows and lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver bromide
7785-23-1
Crystalline
99.998% (metals basis)
MFCD00003398
soluble in alkali cyanide solutions. Partially soluble in ammonia. Insoluble in alcohol and acids. Practically insoluble in water.
Br[Ag]
187.772
187.78
Light sensitive
6.47
432°C
Yellow
25 g
AgBr
silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion
ADZWSOLPGZMUMY-UHFFFAOYSA-M
bromosilver
66199
Odorless
Pale yellow powder/flake/crystalline/beads

Chemical Identifiers

7785-23-1
187.772
ADZWSOLPGZMUMY-UHFFFAOYSA-M
66199
Br[Ag]
AgBr
MFCD00003398
silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion
bromosilver
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.

H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 232-076-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only