missing translation for 'onlineSavingsMsg'
Learn More

Silver bromide, Premion™, 99.998% (metals basis)

Catalog No. p-4782120
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1211006 5 g
AA1211014 25 g
AA1211022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1211006 Supplier Thermo Scientific Chemicals Supplier No. 01211006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7785-23-1 | AgBr | 187.772 g/mol

Silver bromide is used in photographic film. Used in production of radiograph. It also used in infrared applications as windows and lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7785-23-1
Molecular Formula AgBr
Molecular Weight (g/mol) 187.772
MDL Number MFCD00003398
InChI Key ADZWSOLPGZMUMY-UHFFFAOYSA-M
Synonym silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion
PubChem CID 66199
IUPAC Name bromosilver
SMILES Br[Ag]

Specifications

Melting Point 432°C
Color Yellow
Physical Form Crystalline
Quantity 5 g
UN Number UN3077
Merck Index 14,8506
Solubility Information soluble in alkali cyanide solutions. Partially soluble in ammonia. Insoluble in alcohol and acids. Practically insoluble in water.
Formula Weight 187.78
Percent Purity 99.998%
Odor Odorless
Sensitivity Light sensitive
Density 6.47
Assay (metals basis)
Chemical Name or Material Silver bromide, Premion™
Show More Show Less
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 232-076-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only