missing translation for 'onlineSavingsMsg'
Learn More

Silver Bromide, Granular, Purified, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1048100GM

 View more versions of this product

Catalog No. 18613119


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7785-23-1
100 g
Purified
1
99%
Amber Glass Bottle

Chemical Identifiers

7785-23-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions