missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Bromide, Granular, Purified, 99%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 25 g |
| Percent Purity | 99% |
| Grade | Purified |
| Packaging | Amber Glass Bottle |