missing translation for 'onlineSavingsMsg'
Learn More

Silver bis(trifluoromethylsulfonyl)imide

Catalog No. p-7080207
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46784MD 250 mg
AA4678403 1 g
AA4678406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA46784MD Supplier Thermo Scientific Chemicals Supplier No. 046784MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 189114-61-2 | C2AgF6NO4S2 | 388.004 g/mol

Silver bis(trifluoromethylsulfonyl)imide is used in the extraction of C5 - C8 linear alfa olefins using its silver salt to form room temperature ionic liquids. Further, it is utilized for the separation of olefins from paraffins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 189114-61-2
Molecular Formula C2AgF6NO4S2
Molecular Weight (g/mol) 388.004
MDL Number MFCD08457644
InChI Key HSYLTRBDKXZSGS-UHFFFAOYSA-N
Synonym sliverbis trifluoromethanesulfonimide
PubChem CID 131875416
IUPAC Name silver;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ag+]

Specifications

Melting Point 249°C
Color Orange to White
Physical Form Solid
Quantity 250 mg
Linear Formula (CF3SO2)2NAg
UN Number UN1759
Solubility Information Soluble in water.
Formula Weight 388
Sensitivity Light and moisture sensitive
Chemical Name or Material Silver bis (trifluoromethylsulfonyl)imide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only