accessibility menu, dialog, popup

UserName

FFFSOONSOOFFFAg

CAS RN 189114-61-2

IUPAC Name: silver(1+) trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: silver(1+) bistriflylimide anion
Molecular Weight (g/mol): 388
Molecular Formula: C2AgF6NO4S2
InChi Key: HSYLTRBDKXZSGS-UHFFFAOYSA-N
SMILES: [Ag+].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)