missing translation for 'onlineSavingsMsg'
Learn More

Silver Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. p-7135984
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08981G 1 g
S08985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08981G Supplier TCI America Supplier No. S08981G
Only null left

Chemical Identifiers

CAS 189114-61-2
Molecular Formula C2AgF6NO4S2
Molecular Weight (g/mol) 388.004
MDL Number MFCD08457644
InChI Key HSYLTRBDKXZSGS-UHFFFAOYSA-N
Synonym sliverbis trifluoromethanesulfonimide
PubChem CID 131875416
IUPAC Name silver;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ag+]

Specifications

Melting Point 249°C
Quantity 1 g
UN Number 1759
Formula Weight 388.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Silver Bis(trifluoromethanesulfonyl)imide
TSCA No
Recommended Storage Freezer