missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver benzoate hydrate
Description
Silver benzoate hydrate along with 7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene (MTBD) is an efficient catalytic system for the reaction of carbon dioxide with various ketones. It was used in the synthesis of triphenyltinbenzoate. It is also used as a catalyst in Wolff rearrangement reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Cream to White or Gray |
| Physical Form | Powder |
| Quantity | 5 g |
| Linear Formula | C6H5CO2Ag·xH2O |
| Solubility Information | Insoluble in water. |
| IUPAC Name | benzoic acid silver |
| Formula Weight | 228.89 (Anhydrous) |
| Sensitivity | Light sensitive |
| Chemical Name or Material | Silver benzoate hydrate |
Safety and Handling
| EINECSNumber | 208-533-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only