accessibility menu, dialog, popup

UserName

CAS RN 532-31-0

IUPAC Name: silver(1+) benzoate
Synonyms: silver benzoate
Molecular Weight (g/mol): 228.98
Molecular Formula: C7H5AgO2
InChi Key: CLDWGXZGFUNWKB-UHFFFAOYSA-M
SMILES: [Ag+].[O-]C(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results