missing translation for 'onlineSavingsMsg'
Learn More

Silver benzoate hydrate

Catalog No. AA1189614
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1189614 25 g
AA1189606 5 g
AA1189622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1189614 Supplier Thermo Scientific Chemicals Supplier No. 01189614

CAS: 532-31-0 | C7H5AgO2 | 228.98 g/mol

Silver benzoate hydrate along with 7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene (MTBD) is an efficient catalytic system for the reaction of carbon dioxide with various ketones. It was used in the synthesis of triphenyltinbenzoate. It is also used as a catalyst in Wolff rearrangement reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 532-31-0
Molecular Formula C7H5AgO2
Molecular Weight (g/mol) 228.98
MDL Number MFCD00013030
InChI Key CLDWGXZGFUNWKB-UHFFFAOYSA-M
Synonym silver benzoate hydrate
PubChem CID 87384783
SMILES [Ag+].[O-]C(=O)C1=CC=CC=C1

Specifications

Color Cream to White or Gray
Physical Form Powder
Quantity 25 g
Linear Formula C6H5CO2Ag·xH2O
Solubility Information Insoluble in water.
IUPAC Name benzoic acid silver
Formula Weight 228.89 (Anhydrous)
Sensitivity Light sensitive
Chemical Name or Material Silver benzoate hydrate
EINECSNumber 208-533-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only