missing translation for 'onlineSavingsMsg'
Learn More

Silver benzoate, 99%, Thermo Scientific™

Chemical Identifiers

CAS 532-31-0
Molecular Formula C7H6AgO2
Molecular Weight (g/mol) 229.99
MDL Number MFCD00013030
InChI Key GQCYXEOBZQXHPO-UHFFFAOYSA-N
Synonym silver benzoate hydrate
PubChem CID 87384783
IUPAC Name benzoic acid;silver;hydrate
SMILES [Ag].OC(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC210770100
SDS View Documents
Thermo Scientific Chemicals
210770100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC210770500
SDS View Documents
Thermo Scientific Chemicals
210770500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

532-31-0
229.99
GQCYXEOBZQXHPO-UHFFFAOYSA-N
87384783
[Ag].OC(=O)C1=CC=CC=C1
C7H6AgO2
MFCD00013030
silver benzoate hydrate
benzoic acid;silver;hydrate

Specifications

532-31-0
99%
C7H6AgO2
MFCD00013030
15, 1093
Solubility in water: 0.26%. Other solubilities: slightly soluble in alcohol
[Ag].OC(=O)C1=CC=CC=C1
229.99
228.99
Silver benzoate, 99%
Authentic
Glass bottle
C6H5CO2Ag
10g
silver benzoate hydrate
GQCYXEOBZQXHPO-UHFFFAOYSA-N
benzoic acid;silver;hydrate
87384783
99%
Videos
Safety and Handling

Safety and Handling

LIGHT SENSITIVE

EINECSNumber : 208-533-2

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only