missing translation for 'onlineSavingsMsg'
Learn More

Silver Acetate, Powder, Purified, 99%, Spectrum™ Chemical

Catalog No. p-7166267
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
Catalog No. Quantity Packaging
18613115 25 g Amber Glass Bottle
18613116 100 g Amber Glass Bottle
2 options

Catalog No. 18613115

Supplier: Spectrum Chemical Mfg Cor S104525GM

Only null left

Chemical Identifiers

CAS 563-63-3

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade Purified
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.