missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Silicone Sealing Mats for Deep Well Plates, 96 Square

Impermeable sealing mats minimize evaporation in a wide range of temperatures in PCR and storage applications. These silicone mats are chemical-resistant, reusable, and provide a high degree of confidence.

Specifications

Color Natural
Solvent Or Alcohol Resistant Alcohol Resistant, Solvent Resistant
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 96
View More Specs

Products 3
Catalog Number Mfr. No. Pre-slit Material Pierceable Price Quantity & Availability  
Catalog Number Mfr. No. Pre-slit Material Pierceable Price Quantity & Availability  
01183297
View Documents Promotion Details
Simport™ Scientific
T11026
No Polyolefin, Rubber No N/A
Only null left
 
01183298
View Documents Promotion Details
Simport™ Scientific
T11055
No Silicone No N/A
Only null left
 
01183299
View Documents Promotion Details
Simport™ Scientific
T11057
Yes Silicone Yes N/A
Only null left
 
Description

Description

  • Effectively seals deep well plates
  • Easy to open and close
  • Prevents the volatilization of a variety of solvents
  • Chemical and heat resistant
  • All pre-slit models allow the pipette tip to enter the hole without damage to the silicone cover
  • Resistant to DMSO and biological buffers
  • Free of RNase, DNase, Pyrogen, and DNA
Specifications

Specifications

Natural
Non-sterile
96
-80°C to +80°C
Certified Quality
Alcohol Resistant, Solvent Resistant
Autoclavable
Yes
Square
Videos
SDS
Documents

Documents