missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Silicone Sealing Mats for Deep Well Plates, 96 Square

Catalog No. 01183299
Change view
Click to view available options
Pre-slit:
No
Yes
Material:
Polyolefin, Rubber
Silicone
Pierceable:
No
Yes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pre-slit Material Pierceable
01183299 Yes Silicone Yes
01183297 No Polyolefin, Rubber No
01183298 No Silicone No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01183299 Supplier Simport™ Scientific Supplier No. T11057
Only null left

Impermeable sealing mats minimize evaporation in a wide range of temperatures in PCR and storage applications. These silicone mats are chemical-resistant, reusable, and provide a high degree of confidence.

  • Effectively seals deep well plates
  • Easy to open and close
  • Prevents the volatilization of a variety of solvents
  • Chemical and heat resistant
  • All pre-slit models allow the pipette tip to enter the hole without damage to the silicone cover
  • Resistant to DMSO and biological buffers
  • Free of RNase, DNase, Pyrogen, and DNA

Specifications

Color Natural
Solvent Or Alcohol Resistant Alcohol Resistant, Solvent Resistant
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 96
Pre-slit Yes
Certified RNase/DNase-free Yes
Pierceable Yes
Temperature Range (Metric) -80°C to +80°C
Well Shape Square
Fit for Purpose Certified Quality
Show More Show Less