missing translation for 'onlineSavingsMsg'
Learn More

Silicone oil, for melting point and boiling point apparatuses

Catalog No. AC163855000
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163855000 500 g Glass bottle
AC163850010 1 kg Glass bottle
AC163850025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC163855000 Supplier Thermo Scientific Chemicals Supplier No. 163855000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 63148-62-9 | (C2H6OSi)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63148-62-9
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00132673
Synonym Poly(dimethylsiloxane)
IUPAC Name Polydimethylsiloxane
SMILES C[Si](C)(-*)O-*

Specifications

Melting Point -50.0°C
Color Colorless
Physical Form Oily Liquid
Quantity 500 g
Flash Point 300°C
Solubility Information Solubility in water: practically insoluble
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4030 to 1.4060
Specific Gravity 0.96
Density 0.9600g/mL
Chemical Name or Material Silicone oil, For melting point and boiling point apparatuses
Show More Show Less

RUO – Research Use Only