missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Silicone oil
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$1846.87 - $1846.87
Specifications
| CAS | 63148-62-9 |
|---|---|
| Density | 0.913 g/mL (at 25°C) |
| Boiling Point | >140°C (0.002 mmHg) |
| Linear Formula | [-Si(CH3)2 O-]n |
| Refractive Index | n20/D 1.403 (literature) |
Description
Silicone oil is low viscosity liquid polymerized siloxanes with organic side chains.Low viscosity silicone oil has been used to prepare cellulose based electrorheological particles for medical devices. The diffusive behavior of spreading of a silicone oil polymer on solid surfaces was studied. It was also used to investigate Marangoni flow, induced by two liquid droplets on a solid surface, exposed in air. Many diverse applications of silicone oil are as heat exchangers, hydrophobic coating of glass, ceramics, textiles and leathers, component of salves.Specifications
| 63148-62-9 | |
| >140°C (0.002 mmHg) | |
| n20/D 1.403 (literature) | |
| PDMS |
| 0.913 g/mL (at 25°C) | |
| [-Si(CH3)2 O-]n | |
| MFCD00132673 |