missing translation for 'onlineSavingsMsg'
Learn More

Silicone Fluid, 100, Spectrum™ Chemical

Catalog No. p-7166574
Change view
Click to view available options
Quantity:
100 g
1 kg
44 lb.
440 lb.
Packaging:
Amber Glass Bottle
Drum
Steel Pail
Catalog No. Quantity Packaging
18613112 100 g Amber Glass Bottle
18613113 44 lb. Steel Pail
18613114 440 lb. Drum
18613111 1 kg Amber Glass Bottle
4 options

Catalog No. 18613112

Supplier: Spectrum Chemical Mfg Cor SI110100GM

Only null left

Chemical Identifiers

CAS 63148-62-9

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Viscosity 95.0 - 105.0 cs
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.