missing translation for 'onlineSavingsMsg'
Learn More

Silicone Fluid, 100, Spectrum™ Chemical

Catalog No. 18613112
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
1 kg
44 lb.
440 lb.
Packaging:
Amber Glass Bottle
Drum
Steel Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613112 100 g Amber Glass Bottle
18613113 44 lb. Steel Pail
18613114 440 lb. Drum
18613111 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Catalog No. 18613112 missing translation for 'mfr' Spectrum Chemical Mfg Cor missing translation for 'supplierNo' SI110100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 63148-62-9

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Viscosity 95.0 - 105.0 cs
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.