missing translation for 'onlineSavingsMsg'
Learn More

Silicon standard solution, for AAS, 1 mg/ml Si in 2% NaOH

Catalog No. AC196291000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196291000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC196291000 Supplier Thermo Scientific Chemicals Supplier No. 196291000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-21-3 | Si | 28.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy

Specifications

Chemical Name or Material Silicon standard solution
Packaging Plastic bottle
Name Note 1 mg/ml Si in 2% NaOH
CAS 7440-21-3
Density 1.0200g/mL
Physical Form Solution
Molecular Formula Si
Concentration 997 to 1003 μg/mL of Si (4+) in 2%NaOH (20°C)
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES [Si]
IUPAC Name silicon
Molecular Weight (g/mol) 28.09
Quantity 100 mL
PubChem CID 5461123
ChEBI CHEBI:27573
Formula Weight 28.09
Grade AAS
Show More Show Less

RUO – Research Use Only