missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon slugs, 9.5mm (0.375 in.) dia. x 9.5mm (0.375 in.) length, 99.9999% (metals basis), Thermo Scientific Chemicals
$868.53 - $868.53
Chemical Identifiers
| CAS | 7440-21-3 |
|---|---|
| Molecular Formula | Si |
| Molecular Weight (g/mol) | 28.09 |
| MDL Number | MFCD00085311 |
| InChI Key | XUIMIQQOPSSXEZ-UHFFFAOYSA-N |
| Synonym | metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 |
| PubChem CID | 5461123 |
| ChEBI | CHEBI:27573 |
| IUPAC Name | silicon |
| SMILES | [Si] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-21-3 | |
| 28.09 | |
| XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
| 5461123 | |
| silicon |
| Si | |
| MFCD00085311 | |
| metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
| CHEBI:27573 | |
| [Si] |
Specifications
| 7440-21-3 | |
| Odorless | |
| Slug | |
| MFCD00085311 | |
| XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
| silicon | |
| 5461123 | |
| 99.9999% | |
| (metals basis) |
| 25 g | |
| ≈1.57g/slug | |
| Si | |
| metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
| [Si] | |
| 28.09 | |
| CHEBI:27573 | |
| Silicon slugs, 9.5mm (0.375 in.) dia. x 9.5mm (0.375 in.) length |
Safety and Handling
EINECSNumber : 231-130-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only