missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon powder, crystalline, -325 mesh, 99.999% (metals basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Quantity | 10 g |
| Mesh Size | −325 Mesh |
| Odor | Odorless |
| Form | Crystalline Powder |
| UN Number | UN1346 |
| Percent Purity | 99.999% |
| Assay | (metals basis) |
| Chemical Name or Material | Silicon powder |
Safety and Handling
| missing translation for 'healthHazard1' | H228 |
| missing translation for 'healthHazard2' | GHS H Statement H228 Flammable solid. |
| missing translation for 'healthHazard3' | P210-P240-P241-P280-P370+P378q |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: SILICON POWDER, AMORPHOUS |
| EINECSNumber | 231-130-8 |
| RTECSNumber | VW0400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only