missing translation for 'onlineSavingsMsg'
Learn More

Silicon lump, 15cm (5.9in) & down, 99.999+% (metals basis)

CAS: 7440-21-3 | Si | 28.09 g/mol

Supplier:  Thermo Scientific Chemicals 039101A1

 View more versions of this product

Catalog No. AA39101A1

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silicon lump
1 kg
15cm (5.9 in.) & down
(metals basis)
MFCD00085311
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
silicon
5461123
≥99.999%
7440-21-3
Odorless
Lump
Si
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
[Si]
28.09
CHEBI:27573

Chemical Identifiers

7440-21-3
28.09
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
5461123
silicon
Si
MFCD00085311
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
CHEBI:27573
[Si]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-130-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only