missing translation for 'onlineSavingsMsg'
Learn More

Silicon lump, 15cm (5.9in) & down, 99.999+% (metals basis)

Catalog No. AA3910114
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3910114 25 g
AA3910122 100 g
AA39101A1 1 kg
AA39101A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA3910114 Supplier Thermo Scientific Chemicals Supplier No. 03910114

CAS: 7440-21-3 | Si | 28.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]

Specifications

Quantity 25 g
Odor Odorless
Particle Size 15cm (5.9 in.) & down
Form Lump
Percent Purity ≥99.999%
Assay (metals basis)
Chemical Name or Material Silicon lump
EINECSNumber 231-130-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only