Learn More
Silicon granular, virgin poly fines, 99.999% (metals basis)
CAS: 7440-21-3 | Si | 28.09 g/mol
Supplier: Thermo Scientific Chemicals 03854222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silicon granular | |
| 7440-21-3 | |
| Odorless | |
| Chunks to Powder | |
| Si | |
| metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
| [Si] | |
| 28.09 | |
| CHEBI:27573 |
| Virgin poly fines | |
| 100 g | |
| ∽1/4 in. & down | |
| (metals basis) | |
| MFCD00085311 | |
| XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
| silicon | |
| 5461123 | |
| 99.999% |
Chemical Identifiers
| 7440-21-3 | |
| 28.09 | |
| XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
| 5461123 | |
| silicon |
| Si | |
| MFCD00085311 | |
| metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
| CHEBI:27573 | |
| [Si] |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
EINECSNumber : 231-130-8
RTECSNumber : VW0400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only