missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon Dioxide, 325 Mesh, FCC, 90.4%, Spectrum™ Chemical
S1388, 7631-86-9, SiO2·xH2O
$846.83 - $4810.27
Chemical Identifiers
| CAS | 7631-86-9 |
|---|---|
| Molecular Formula | O2Si |
| Molecular Weight (g/mol) | 60.08 |
| MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
| InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
| IUPAC Name | silanedione |
| SMILES | O=[Si]=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606217
|
Spectrum Chemical Mfg Cor
S138825KG |
2.5 kg | Poly Pail |
Each for $846.83
|
|
||||
|
18606218
|
Spectrum Chemical Mfg Cor
S138812KG |
12 kg | Fiber Drum |
Each for $2,083.47
|
|
||||
|
18606219
|
Spectrum Chemical Mfg Cor
S138820KGBL |
20 kg | Fiber Drum |
Each for $4,810.27
|
|
||||
Description
Spectrum™ Chemical Silicon Dioxide, 325 Mesh, FCC is used as a food additive as a powder flow helper. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chemical Identifiers
| 7631-86-9 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| O=[Si]=O |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| silanedione |
Specifications
| 7631-86-9 | |
| Silicon Dioxide | |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| 325 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione | |
| 94% | |
| Silicon Dioxide |
| 1 | |
| 2.5 kg | |
| 0.07 | |
| 0.085 | |
| Poly Pail | |
| O=[Si]=O | |
| 60.08 | |
| FCC |