missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon Dioxide, 325 Mesh, FCC, 90.4%, Spectrum™ Chemical
S1388, 7631-86-9, SiO2·xH2O
Supplier: Spectrum Chemical Mfg Cor S138812KG
Description
Spectrum™ Chemical Silicon Dioxide, 325 Mesh, FCC is used as a food additive as a powder flow helper. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 7631-86-9 | |
| 12 kg | |
| 0.07 | |
| 0.085 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione | |
| 94% |
| 1 | |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| Fiber Drum | |
| O=[Si]=O | |
| 60.08 | |
| FCC |
Chemical Identifiers
| 7631-86-9 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| O=[Si]=O |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| silanedione |