missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon AA Standard, Certified, 1000ppm Si ±10ppm (1mL = 1mg Si), LabChem™
$75.37 - $152.60
Chemical Identifiers
| CAS | 13517-24-3 |
|---|---|
| Molecular Formula | H18Na2O12Si |
| Molecular Weight (g/mol) | 284.197 |
| InChI Key | PHIQPXBZDGYJOG-UHFFFAOYSA-N |
| PubChem CID | 61639 |
| IUPAC Name | disodium;dioxido(oxo)silane;nonahydrate |
| SMILES | O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+] |
Chemical Identifiers
| 13517-24-3 | |
| 284.197 | |
| 61639 | |
| O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+] |
| H18Na2O12Si | |
| PHIQPXBZDGYJOG-UHFFFAOYSA-N | |
| disodium;dioxido(oxo)silane;nonahydrate |
Specifications
| Poly Bottle | |
| 98.99,1.01 | |
| 1g/mL | |
| H18Na2O12Si | |
| Na2SiO3·9H2O | |
| Soluble in water | |
| O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+] | |
| 284.197 | |
| 61639 | |
| Certified |
| 13517-24-3,7732-18-5 | |
| Passes Test | |
| Liquid | |
| 1000 ppm Si ±10 ppm (1mL = 1mg Si) | |
| Colorless | |
| PHIQPXBZDGYJOG-UHFFFAOYSA-N | |
| disodium;dioxido(oxo)silane;nonahydrate | |
| 125 mL | |
| 284.2 | |
| Silicon AA Standard, 1000ppm (1mL = 1mg Si) |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
Recommended Storage : Room Temperature