missing translation for 'onlineSavingsMsg'
Learn More

Silicic acid hydrate

Catalog No. AA3327022
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3327022 100 g
AA3327036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3327022 Supplier Thermo Scientific Chemicals Supplier No. 03327022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1343-98-2 | O2Si | 60.08 g/mol

Silicic acid hydrate is used for studying neurodegenerative diseases due to its ability to inhibit aluminum uptake. It is utilized for the manufacture catalyst and catalyst carrier. It is involved in tungsten filament production as chemical reagent and flux. Further, it is used for oil and wax decoloring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1343-98-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym orthosilicic acid, monosilicic acid, tetrahydroxysilane, h4sio4, silicon tetrahydroxide, silicasol, kieselsaeure, si oh 4, unii-623b93yabh, orthosilicicacid
PubChem CID 14942
ChEBI CHEBI:26675
IUPAC Name silicic acid
SMILES O=[Si]=O

Specifications

Appearance White powder
Color White
Quantity 100 g
Physical Form -100 Mesh Powder
Formula Weight 60.09 (Anhydrous)
Chemical Name or Material Silicic acid hydrate
Hazard Category H373: May cause damage to organs through prolonged or repeated exposure.
EINECSNumber 215-683-2
RTECSNumber VV8853000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only